Learn More
Eurofins Discoverx PP5, active, 250 µg. (14-778M)
Supplier:  Eurofins Discoverx 14778M
Encompass
The PP5, active product is purified N terminal GST-tagged recombinant PP5 enzyme. This enzyme is full length. It retains its binding and enzymatic activity and is intended for use in phosphatase assays. This enzyme is provided as 250 μg of 1.453 mg/mL frozen solution. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9328