Learn More
Enzo Life Sciences pp60 v-src autophosphorylation site (1mg). CAS: 81493-98-3
SDP
Supplier:  Enzo Life Sciences BMLP3070001
Encompass_Preferred
Residues 2-12 are the v-Src (412-422) autophosphorylation site with an extra Arg for phosphocellulose binding. This substrate sequence, RRLIEDNEYTARG, was originally described as an EGFR substrate in A431 cell extracts (Km=0.3-0.6 mM). Purity: ≥97%. Long Term Storage: -20°C.
Catalog No. 50-201-1338