Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PPA | 452072-22-9 | 99.0% | 243.35 | C10H13NO2S2 | 25 MG
SDP

Catalog No. 5000202892
Encompass_Preferred

PPA is an ADC linker used to prepare antibody-drug conjugates, referenced by patent US20060073528A1. It is supplied with high purity and is available in multiple pack sizes; technical documents such as a data sheet, certificate of analysis, and safety data sheet are provided.

  • Used for antibody-drug conjugate synthesis.
  • Purity 99.01%.
  • CAS number 452072-22-9.
  • Available in 25 mg, 50 mg, 100 mg, 200 mg, 500 mg pack sizes.
  • Technical documents: data sheet, COA, SDS available.

Catalog No. 50-002-02892 Supplier Medchemexpress LLC Supplier No. HY14166425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PPA is an ADC linker used to prepare antibody-drug conjugates, referenced by patent US20060073528A1. It is supplied with high purity and is available in multiple pack sizes; technical documents such as a data sheet, certificate of analysis, and safety data sheet are provided.

  • Used for antibody-drug conjugate synthesis.
  • Purity 99.01%.
  • CAS number 452072-22-9.
  • Available in 25 mg, 50 mg, 100 mg, 200 mg, 500 mg pack sizes.
  • Technical documents: data sheet, COA, SDS available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.