Learn More
TARGETMOL CHEMICALS INC PPA-904 10MG
SDP
Catalog No. 502878379
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. PPA-904, a specific phenothiazine photosensitizer, is used in photodynamic therapy (PDT). Purity 99.2%
Catalog No. 50-287-8379 Supplier TARGETMOL CHEMICALS INC Supplier No. T1952510MG
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. PPA-904, a specific phenothiazine photosensitizer, is used in photodynamic therapy (PDT). Purity 99.2%