Learn More
Medchemexpress LLC PPA-904 | 30189-85-6 | 532.62 | 1 ML
SDP
Catalog No. 5000446903
Encompass_Preferred
PPA-904 is a specific phenothiazine photosensitizer used in photodynamic research (PDT), particularly for topical application in cutaneous leishmaniasis in vivo. PPA-904 PDT has the potential to provide an immunomodulating, dose-dependent efflux on IL-12p70 production, which may contribute to a more rapid healing in treated mice.
  • Specific phenothiazine photosensitizer
  • Used in photodynamic therapy (PDT) research
  • Applied topically for cutaneous leishmaniasis studies in vivo
  • Targets parasites like Leishmania
  • Induces immunomodulating, dose-dependent efflux on IL-12p70 production
  • May contribute to more rapid healing in treated mice
Catalog No. 50-004-46903 Supplier Medchemexpress LLC Supplier No. HYU0012810MM/1ML
PPA-904 is a specific phenothiazine photosensitizer used in photodynamic research (PDT), particularly for topical application in cutaneous leishmaniasis in vivo. PPA-904 PDT has the potential to provide an immunomodulating, dose-dependent efflux on IL-12p70 production, which may contribute to a more rapid healing in treated mice.
  • Specific phenothiazine photosensitizer
  • Used in photodynamic therapy (PDT) research
  • Applied topically for cutaneous leishmaniasis studies in vivo
  • Targets parasites like Leishmania
  • Induces immunomodulating, dose-dependent efflux on IL-12p70 production
  • May contribute to more rapid healing in treated mice