Learn More
Medchemexpress LLC PPA-904 | 30189-85-6 | 532.62 | 100 MG
SDP
Catalog No. 5000449644
Encompass_Preferred
PPA-904 is a specific phenothiazine photosensitizer utilized in photodynamic research (PDT), particularly for topical application in cutaneous leishmaniasis in vivo. It is intended for research use only and not sold to patients.
  • Specific phenothiazine photosensitizer
  • Used in photodynamic research (PDT)
  • For topical application in cutaneous leishmaniasis in vivo
  • Research use only
  • High purity of 99.85%
  • Solid appearance, brown to black color
  • Targets Leishmania
  • Induces immunomodulating, dose-dependent efflux on IL-12p70 production
Catalog No. 50-004-49644 Supplier Medchemexpress LLC Supplier No. HYU00128100MG
PPA-904 is a specific phenothiazine photosensitizer utilized in photodynamic research (PDT), particularly for topical application in cutaneous leishmaniasis in vivo. It is intended for research use only and not sold to patients.
  • Specific phenothiazine photosensitizer
  • Used in photodynamic research (PDT)
  • For topical application in cutaneous leishmaniasis in vivo
  • Research use only
  • High purity of 99.85%
  • Solid appearance, brown to black color
  • Targets Leishmania
  • Induces immunomodulating, dose-dependent efflux on IL-12p70 production