Learn More
Medchemexpress LLC PPA-904 | 30189-85-6 | 532.62 | 5 MG
SDP
Catalog No. 5000448559
Encompass_Preferred
PPA-904 is a specific phenothiazine photosensitizer used in photodynamic research (PDT), particularly for topical application in cutaneous leishmaniasis in vivo.
  • PPA904-PDT can provide an immunomodulating, dose-dependent effect on IL-12p70 production, which may promote more rapid healing in PPA904-PDT treated mice.
  • The IC50 target is Leishmania.
Catalog No. 50-004-48559 Supplier Medchemexpress LLC Supplier No. HYU001285MG
PPA-904 is a specific phenothiazine photosensitizer used in photodynamic research (PDT), particularly for topical application in cutaneous leishmaniasis in vivo.
  • PPA904-PDT can provide an immunomodulating, dose-dependent effect on IL-12p70 production, which may promote more rapid healing in PPA904-PDT treated mice.
  • The IC50 target is Leishmania.