Learn More
Aviva Systems Biology Corp PPA1 Antibody - N-terminal region (ARP48396_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48396_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PPA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PPA1. Alternate names: IOPPP, MGC111556, PP, PP1, SID6-8061ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-59812