Learn More
Enzo Life Sciences PPACK (25mg). CAS: 71142-71-7
SDP
Supplier:  Enzo Life Sciences BMLPI1170025
Encompass_Preferred
Potent irreversible thrombin inhibitor. Inhibits thrombin activation of progelatinase A. Pretreatment with PPACK inhibits thrombin-induced cerebral thromboembolism in vivo. Alternative name: D-Phe-Pro-Arg-CMK. Purity: ≥90% (HPLC). Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-1457