Learn More
Haematologic Technologies Inc PPACK 5MG
Catalog No. 50883455
Encompass_Preferred
Inhibitor, Phe-Pro-Arg Chloromethylketone (PPACK), 5 milligrams, Use: Tri-peptide chloromethylketones are used to irreversibly inhibit various serine proteases, Research use only, Supplied lyophilized, store at 4 degrees Celsius.
Catalog No. 50-883-455 Supplier Haematologic Technologies Inc Supplier No. FPRCK015MG
Inhibitor, Phe-Pro-Arg Chloromethylketone (PPACK), 5 milligrams, Use: Tri-peptide chloromethylketones are used to irreversibly inhibit various serine proteases, Research use only, Supplied lyophilized, store at 4 degrees Celsius.