Learn More
Apexbio Technology LLC PPACK Dihydrochloride 142036-63-3 5mg
SDP
Catalog No. 502240378
Encompass_Preferred
PPACK Dihydrochloride is a small-molecule inhibitor targeting human -thrombin It is designed to inhibit thrombin activity thereby blocking downstream platelet aggregation and coagulation pathways PPACK Dihydrochloride exerts its biological activity primarily through selective irreversible inhibition of thrombin by covalently occupying the thrombin active-site serine residue and forming a stable tetrahedral intermediate with the catalytic His57 residue Based on these pharmacological properties PPACK Dihydrochloride holds research potential in biomedical studies as an anticoagulant reagent and for investigating thrombin-mediated coagulation and platelet activation mechanisms
This item is not returnable. View return policy
Catalog No. 50-224-0378 Supplier Apexbio Technology LLC Supplier No. A25885
PPACK Dihydrochloride is a small-molecule inhibitor targeting human -thrombin It is designed to inhibit thrombin activity thereby blocking downstream platelet aggregation and coagulation pathways PPACK Dihydrochloride exerts its biological activity primarily through selective irreversible inhibition of thrombin by covalently occupying the thrombin active-site serine residue and forming a stable tetrahedral intermediate with the catalytic His57 residue Based on these pharmacological properties PPACK Dihydrochloride holds research potential in biomedical studies as an anticoagulant reagent and for investigating thrombin-mediated coagulation and platelet activation mechanisms