Learn More
Medchemexpress LLC Ppack TFA | 157379-44-7 | 99.8% | 5 MG
SDP
Catalog No. 5000232025
Encompass_Preferred
PPACK TFA is a potent, selective, and irreversible thrombin inhibitor supplied as the trifluoroacetate (TFA) salt. It is used as an anticoagulant alternative for blood gas and whole-blood electrolyte analyses and inhibits plasminogen activator (rt-PA) binding to plasma protease inhibitors.
  • Irreversible thrombin inhibitor for anticoagulant applications.
  • Supplied as the trifluoroacetate salt for stability and handling.
  • High analytical purity (99.8%).
  • Suitable for blood gas and whole-blood electrolyte analyses.
  • Available in a 5 mg research-size vial.
  • CAS number: 157379-44-7.
Catalog No. 50-002-32025 Supplier Medchemexpress LLC Supplier No. HY122542B5MG
PPACK TFA is a potent, selective, and irreversible thrombin inhibitor supplied as the trifluoroacetate (TFA) salt. It is used as an anticoagulant alternative for blood gas and whole-blood electrolyte analyses and inhibits plasminogen activator (rt-PA) binding to plasma protease inhibitors.
  • Irreversible thrombin inhibitor for anticoagulant applications.
  • Supplied as the trifluoroacetate salt for stability and handling.
  • High analytical purity (99.8%).
  • Suitable for blood gas and whole-blood electrolyte analyses.
  • Available in a 5 mg research-size vial.
  • CAS number: 157379-44-7.