Learn More
System Biosciences LLC pPACKH1 HIV LENTIVECTOR PACKAGING KIT - 10TpPACKH1 HIV Lentivector Packaging Kit= 10 Transfections
SDP
Catalog No. NC9985269
Encompass_Preferred
Generate high titers of transduction-ready pseudoviral particles with this HIV-based Lentivector Packaging Kit. Designed to efficiently package most third-generation lentivectors, the pPACKH1 HIV Lentivector Packaging Kit makes it easy to deliver and express lentiviral constructs in a wide-range of mammalian cells.
Catalog No. NC9985269 Supplier System Biosciences LLC Supplier No. LV500A1
Generate high titers of transduction-ready pseudoviral particles with this HIV-based Lentivector Packaging Kit. Designed to efficiently package most third-generation lentivectors, the pPACKH1 HIV Lentivector Packaging Kit makes it easy to deliver and express lentiviral constructs in a wide-range of mammalian cells.