Learn More
Cayman Chemical PPACK (hydrochloride), Purity: ≥95%, CAS: 82188-90-7, 10 mg
SDP
Catalog No. 502917858
Encompass_Preferred
A synthetic peptide derivative that irreversibly and specifically inhibits thrombin-mediated platelet activation by binding with high affinity to the active site of thrombin (Ki = 0.24 nM); used as an anticoagulant (100 μM)
Catalog No. 50-291-7858 Supplier Cayman Chemical Supplier No. 3748910 MG
A synthetic peptide derivative that irreversibly and specifically inhibits thrombin-mediated platelet activation by binding with high affinity to the active site of thrombin (Ki = 0.24 nM); used as an anticoagulant (100 μM)