Learn More
Cayman Chemical PPACKtrIfluoroacetAT salt 10mg
SDP
Catalog No. 502971656
Encompass_Preferred
A synthetic peptide derivative that irreversibly and specifically inhibits thrombin-mediated platelet activation by binding with high affinity to the active site of thrombin (Ki = 0.24 nM); used as an anticoagulant (100 μM)
Catalog No. 50-297-1656 Supplier Cayman Chemical Supplier No. 1516010 MG
A synthetic peptide derivative that irreversibly and specifically inhibits thrombin-mediated platelet activation by binding with high affinity to the active site of thrombin (Ki = 0.24 nM); used as an anticoagulant (100 μM)