Learn More
Cayman Chemical PPADS (sodium salt), Purity: ≥95%, CAS: 192575-19-2, 10 mg
SDP
Catalog No. 502969393
Encompass_Preferred
A non-selective P2 purinergic antagonist that blocks recombinant P2X1-5 (IC50s = 1-2.6 μM), native P2Y2-like (IC50 = ~0.9 mM), and recombinant P2Y4 (IC50 = ~15 mM) receptors; produces neuroprotective effects in an ischemia model with permanent middle cerebral artery occlusion in spontaneously hypertensive rats
Catalog No. 50-296-9393 Supplier Cayman Chemical Supplier No. 1453710 MG
A non-selective P2 purinergic antagonist that blocks recombinant P2X1-5 (IC50s = 1-2.6 μM), native P2Y2-like (IC50 = ~0.9 mM), and recombinant P2Y4 (IC50 = ~15 mM) receptors; produces neuroprotective effects in an ischemia model with permanent middle cerebral artery occlusion in spontaneously hypertensive rats