Learn More
Cayman Chemical PPAdodIum salt 50mg
SDP
Catalog No. 502970678
Encompass_Preferred
A non-selective P2 purinergic antagonist that blocks recombinant P2X1-5 (IC50s = 1-2.6 μM), native P2Y2-like (IC50 = ~0.9 mM), and recombinant P2Y4 (IC50 = ~15 mM) receptors; produces neuroprotective effects in an ischemia model with permanent middle cerebral artery occlusion in spontaneously hypertensive rats
Catalog No. 50-297-0678 Supplier Cayman Chemical Supplier No. 1453750 MG
A non-selective P2 purinergic antagonist that blocks recombinant P2X1-5 (IC50s = 1-2.6 μM), native P2Y2-like (IC50 = ~0.9 mM), and recombinant P2Y4 (IC50 = ~15 mM) receptors; produces neuroprotective effects in an ischemia model with permanent middle cerebral artery occlusion in spontaneously hypertensive rats