Learn More
Aviva Systems Biology Corp PLPP1 Antibody - N-terminal region (ARP42721_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42721_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PPAP2A. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human PPAP2A. Alternate names: LLP1a, LPP1, PAP-2a, PAP2, PAP2a2, PAP2alpha2, PAPalpha1ApplicationIHC, WB. Species reactivity: Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-58317