Learn More
Aviva Systems Biology Corp PLPP1 Antibody - middle region (ARP42146_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42146_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PPAP2A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PPAP2A. Alternate names: LLP1a, LPP1, PAP-2a, PAP2, PAP2a2, PAP2alpha2, PAPalpha1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53324