Learn More
Aviva Systems Biology Corp PLPP1 Antibody - N-terminal region (ARP42145_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42145_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPAP2A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human PPAP2A. Alternate names: LLP1a, LPP1, PAP-2a, PAP2, PAP2a2, PAP2alpha2, PAPalpha1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-979