Learn More
Aviva Systems Biology Corp PLPP5 Antibody - middle region (ARP50074_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50074_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPAPDC1B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PPAPDC1B. Alternate names: DPPL1, HTPAPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53616