Learn More
Aviva Systems Biology Corp PLPP6 Antibody - N-terminal region (ARP55974_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55974_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPAPDC2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PPAPDC2. Alternate names: FLJ46512, FLJ90191, MGC15483, PSDP, bA6J24.6, PDP1ApplicationWB. Species reactivity: Dog, Human
Catalog No. 50-014-61679