Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PPARγ agonist 5 | 915124-86-6 | 98.59% | 513.63 | 25 MG
SDP

Catalog No. 5000363972
Encompass_Preferred

PPARγ agonist 5 is a potent and selective agonist of PPARγ with potential for research into cancer diseases. It appears as a solid with a white to off-white color.

  • Potent and selective agonist of PPARγ
  • Potential for cancer disease research
  • Solid, white to off-white appearance
  • Purity: 98.59%
  • Molecular weight: 513.63
  • Soluble in DMSO (100 mg/mL)

Catalog No. 50-003-63972 Supplier Medchemexpress LLC Supplier No. 25MGHY146480
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PPARγ agonist 5 is a potent and selective agonist of PPARγ with potential for research into cancer diseases. It appears as a solid with a white to off-white color.

  • Potent and selective agonist of PPARγ
  • Potential for cancer disease research
  • Solid, white to off-white appearance
  • Purity: 98.59%
  • Molecular weight: 513.63
  • Soluble in DMSO (100 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.