Learn More
Enzo Life Sciences PPAR gamma 2 (mouse) polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ALX210192R100
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to aa 1-16 (M1GETLGDSPIDPESDS16) of human PPARγ2 (peroxisome proliferator activated receptor γ2). . Species reactivity: Mouse. Applications: ICC, IHC, WB, FUNC Gel Supershift. Purity: Epitope-affinity purified IgG. Formulation: Liquid. In PBS containing 1mg/ml BSA and 0.05% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-8088