Learn More
Medchemexpress LLC PPAR-gamma Human 2 50ug | 50ug
SDP
Catalog No. 5000194827
Encompass_Preferred
The PPAR gamma protein is a nuclear receptor that binds to peroxisome proliferators and is activated upon ligand binding to specific PPREs on DNA It regulates target gene transcription and controls fatty acid metabolism PPAR-gamma Protein Human (271aa His solution) is the recombinant human-derived PPAR-gamma protein expressed by E coli with labeled tag The total length of PPAR-gamma Protein Human (271aa His solution) is 271 a a with molecular weight of 31 kDa
Catalog No. 50-001-94827 Supplier Medchemexpress LLC Supplier No. HYP790012Y50UG
The PPAR gamma protein is a nuclear receptor that binds to peroxisome proliferators and is activated upon ligand binding to specific PPREs on DNA It regulates target gene transcription and controls fatty acid metabolism PPAR-gamma Protein Human (271aa His solution) is the recombinant human-derived PPAR-gamma protein expressed by E coli with labeled tag The total length of PPAR-gamma Protein Human (271aa His solution) is 271 a a with molecular weight of 31 kDa