Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ppar gamma protein, human (C-His, solution) | >90.0% | 50UG
SDP

Catalog No. 5000177462
Encompass_Preferred

Recombinant fragment of human peroxisome proliferator-activated receptor gamma representing the ligand-binding domain (residues D238-D503). Supplied as a C-terminal His-tagged solution expressed in E. coli and formulated for biochemical and biophysical assays, this product offers high purity and low endotoxin suitable for binding, screening, and structural studies.

  • Recombinant human PPARγ ligand-binding domain (residues D238-D503).
  • C-terminal His tag for detection and purification.
  • Expressed in E. coli and supplied as a 0.22 μm filtered solution.
  • Formulated in 50 mM Tris, 300 mM NaCl, 500 mM arginine, pH 8.0, 10% glycerol.
  • Approximate molecular weight 30 kDa.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Endotoxin level <1 EU/μg (LAL method).
  • Suitable for biochemical binding, ligand screening, and structural studies.

Catalog No. 50-001-77462 Supplier Medchemexpress LLC Supplier No. HYP70027550UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant fragment of human peroxisome proliferator-activated receptor gamma representing the ligand-binding domain (residues D238-D503). Supplied as a C-terminal His-tagged solution expressed in E. coli and formulated for biochemical and biophysical assays, this product offers high purity and low endotoxin suitable for binding, screening, and structural studies.

  • Recombinant human PPARγ ligand-binding domain (residues D238-D503).
  • C-terminal His tag for detection and purification.
  • Expressed in E. coli and supplied as a 0.22 μm filtered solution.
  • Formulated in 50 mM Tris, 300 mM NaCl, 500 mM arginine, pH 8.0, 10% glycerol.
  • Approximate molecular weight 30 kDa.
  • Purity greater than 90% by reducing SDS-PAGE.
  • Endotoxin level <1 EU/μg (LAL method).
  • Suitable for biochemical binding, ligand screening, and structural studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.