Learn More
Medchemexpress LLC PPAR gamma Human H 100ug | 100ug
SDP
Catalog No. 5000194643
Encompass_Preferred
PPAR gamma LBD Protein Human (His) is a His-fused PPAR-gamma-LBD protein expressed by E coli approximately 32 6 kDa PPAR gamma LBD Protein can be used in the ligand screening assays western blotting and ELISA et al[1]
Catalog No. 50-001-94643 Supplier Medchemexpress LLC Supplier No. HYP7999100UG
PPAR gamma LBD Protein Human (His) is a His-fused PPAR-gamma-LBD protein expressed by E coli approximately 32 6 kDa PPAR gamma LBD Protein can be used in the ligand screening assays western blotting and ELISA et al[1]