Learn More
Medchemexpress LLC PPAR gamma Human s 10ug
SDP
Catalog No. 5000179500
Encompass_Preferred
The PPAR gamma protein is a nuclear receptor that binds to peroxisome proliferators and is activated upon ligand binding to specific PPREs on DNA It regulates target gene transcription and controls fatty acid metabolism PPAR gamma Protein Human (sf9 His-GST) is the recombinant human-derived PPAR gamma protein expressed by Sf9 insect cells with N-His N-GST labeled tag
Catalog No. 50-001-79500 Supplier Medchemexpress LLC Supplier No. HYP7370110UG
The PPAR gamma protein is a nuclear receptor that binds to peroxisome proliferators and is activated upon ligand binding to specific PPREs on DNA It regulates target gene transcription and controls fatty acid metabolism PPAR gamma Protein Human (sf9 His-GST) is the recombinant human-derived PPAR gamma protein expressed by Sf9 insect cells with N-His N-GST labeled tag