Learn More
Medchemexpress LLC PPAR gamma Human s 50ug
SDP
Catalog No. 5000179452
Encompass_Preferred
The PPAR gamma protein is a nuclear receptor that binds to peroxisome proliferators and is activated upon ligand binding to specific PPREs on DNA It regulates target gene transcription and controls fatty acid metabolism PPAR gamma Protein Human (sf9 His-GST) is the recombinant human-derived PPAR gamma protein expressed by Sf9 insect cells with N-His N-GST labeled tag
Catalog No. 50-001-79452 Supplier Medchemexpress LLC Supplier No. HYP7370150UG
The PPAR gamma protein is a nuclear receptor that binds to peroxisome proliferators and is activated upon ligand binding to specific PPREs on DNA It regulates target gene transcription and controls fatty acid metabolism PPAR gamma Protein Human (sf9 His-GST) is the recombinant human-derived PPAR gamma protein expressed by Sf9 insect cells with N-His N-GST labeled tag