Learn More
Cayman Chemical PPAR2regIonAB MoncLNal100uL
SDP
Catalog No. 502948603
Encompass_Preferred
An internal standard for the quantification of hydroflumethiazide by GC- or LC-MS
Catalog No. 50-294-8603 Supplier Cayman Chemical Supplier No. 32805100 UL
An internal standard for the quantification of hydroflumethiazide by GC- or LC-MS