Learn More
Aviva Systems Biology Corp PPARA Antibody - middle region (ARP34113_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34113_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PPARA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PPARA. Alternate names: PPAR, NR1C1, hPPAR, PPARalphaApplicationWB. Species reactivity: Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-55252