Learn More
Aviva Systems Biology Corp PPARA Antibody - N-terminal region (ARP33066_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33066_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPARA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the n terminal region of human PPARA. Alternate names: MGC2237, MGC2452, NR1C1, PPAR, hPPAR, PPARalphaApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54868