Learn More
Aviva Systems Biology Corp PPARA Antibody - middle region (ARP34112_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34112_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPARA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PPARA. Alternate names: MGC2237, MGC2452, NR1C1, PPAR, hPPAR, anon-EST:Liang-1.32, clone 1.32, Dmel\CG8486, PPARAApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Mouse, Pig, Rat, Sheep
Catalog No. 50-014-55251