Learn More
Cayman Chemical PPARα Transcription Factor Assay Kit, 96 Well
SDP
Catalog No. 502886994
Encompass_Preferred
PPARs are ligand-activated transcription factors belonging to the large superfamily of nuclear receptors. PPARα primarily activates genes encoding proteins involved in fatty acid oxidation, while PPARγ primarily activates genes directly involved in lipogenic pathway and insulin signalling. Members of the PPAR family are important direct targets of many antidiabetic and hypolipidemic drugs. Cayman’s PPAR Transcription Factor Assays are a non-radioactive, sensitive method for detecting specific transcription factor DNA binding activity in nuclear extracts and whole cell lysates.
Catalog No. 50-288-6994 Supplier Cayman Chemical Supplier No. 1000691596 WELL
PPARs are ligand-activated transcription factors belonging to the large superfamily of nuclear receptors. PPARα primarily activates genes encoding proteins involved in fatty acid oxidation, while PPARγ primarily activates genes directly involved in lipogenic pathway and insulin signalling. Members of the PPAR family are important direct targets of many antidiabetic and hypolipidemic drugs. Cayman’s PPAR Transcription Factor Assays are a non-radioactive, sensitive method for detecting specific transcription factor DNA binding activity in nuclear extracts and whole cell lysates.