Learn More
Enzo Life Sciences PPARalpha monoclonal antibody (3B6/PPAR) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804255C100
Encompass_Preferred
Clone: 3B6/PPAR. Isotype: IgG2b. Immunogen: Purified recombinant PPARα (peroxisome proliferator activated receptor α). Species reactivity: Human, Mouse, Rat. Applications: IP, WB Gel Shift. Purity: Purified IgG2b. Formulation: Liquid. In PBS containing 0.05% sodium azide.
Catalog No. 50-200-9843