Learn More
Aviva Systems Biology Corp PPARD Antibody - middle region (ARP32878_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32878_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PPARD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PPARD. Alternate names: FAAR, NUC1, NUCI, NR1C2, NUCII, PPARBApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54803