Learn More
Aviva Systems Biology Corp PPARD Antibody - N-terminal region (ARP37889_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37889_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PPARD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of mouse PPARD. Alternate names: NUC1, NUC-1, Nr1c2, Pparb, PPAR[b], Pparb/d, PPAR-beta, PPARdelta, PPAR-deltaApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-56457