Learn More
Aviva Systems Biology Corp PPARG Antibody - N-terminal region (ARP39207_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39207_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PPARG. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PPARG. Alternate names: GLM1, CIMT1, NR1C3, PPARG1, PPARG2, PPARgammaApplicationWB. Species reactivity: Dog, Human, Pig
Catalog No. 50-014-56942