Learn More
Aviva Systems Biology Corp PPARG Antibody - N-terminal region (ARP32880_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32880_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PPARG. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PPARG. Alternate names: GLM1, CIMT1, NR1C3, PPARG1, PPARG2, PPARgammaApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-54805