Learn More
Aviva Systems Biology Corp PPARGC1A Antibody - N-terminal region (ARP39015_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39015_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPARGC1A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human PPARGC1A. Alternate names: LEM6, PGC-1(alpha), PGC-1v, PGC1, PGC1A, PPARGC1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-774