Learn More
Aviva Systems Biology Corp Ppargc1a Antibody - C-terminal region (ARP39016_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39016_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ppargc1a. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: A830037N07Rik, PGC-1, PGC-1v, Pgc-1alpha, Pgc1, Pgco1, Ppargc1, Gm11133, ENSMUSG00000079510ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-775