Learn More
Cayman Chemical PPARLBD LIgnd Screeng384
SDP
Catalog No. 502895846
Encompass_Preferred
A growth hormone secretagogue and ghrelin receptor agonist binds to GHS-R1a (IC50 123 nM for the human receptor) oral administration increases plasma growth hormone levels in dogs at 1 mg/kg decreases gastric acid secretion in pylorus-ligated rats at 0.01 or 1 nmol/animal i.c.v. increases acute food intake and body weight in mice at 10 mg/kg decreases the number and total duration of seizures in mice induced with dopamine D1 receptor kindling using SKF 81297
Catalog No. 50-289-5846 Supplier Cayman Chemical Supplier No. 600616384 WELL
A growth hormone secretagogue and ghrelin receptor agonist binds to GHS-R1a (IC50 123 nM for the human receptor) oral administration increases plasma growth hormone levels in dogs at 1 mg/kg decreases gastric acid secretion in pylorus-ligated rats at 0.01 or 1 nmol/animal i.c.v. increases acute food intake and body weight in mice at 10 mg/kg decreases the number and total duration of seizures in mice induced with dopamine D1 receptor kindling using SKF 81297