Learn More
Echelon Biosciences Research Labs XY-4 (PPAR agonist) 0.5 mg
SDP
Supplier:  Echelon Biosciences Research Labs L1160500UG
Encompass
(2R)-1,1-difluoro-2-palmitoyloxy-3-phosphopropane (XY-4) is an analog of 2-palmitoyl lysophosphatidic acid (LPA) in which the sn-1 hydroxyl has been replaced by two fluorines. It acts as a selective agonist of peroxisome proliferator-activated receptor (PPAR) but does not stimulate LPA1, LPA2, or LPA3 receptors. PPAR is a transcription factor of the nuclear receptor superfamily and has been implicated in atherogenesis. This item is non returnable
Catalog No. 50-720-493