Learn More
Trajan Scientific Pure-Pass 96 well plates, polypropylene, V bottom, round, 7 mm, 1.0 mL, PK10
SDP
Catalog No. 502723961
Encompass_Preferred
Pure-Pass 96 well plates, polypropylene, V bottom, round, 7 mm, 1.0 mL, PK10
Catalog No. 50-272-3961 Supplier Trajan Scientific Supplier No. 451002
Add to Cart
Add to Cart
Pure-Pass 96 well plates, polypropylene, V bottom, round, 7 mm, 1.0 mL, PK10