Learn More
Aviva Systems Biology Corp PPAT Antibody - C-terminal region (ARP46080_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46080_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPAT. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PPAT. Alternate names: ATASE, GPAT, PRATApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Rat
Catalog No. 50-014-59214