Learn More
MOLTOX Molecular Toxicology Inc Phosphate Peptone Buffer (PPB), pH 7.0 +/- 0.2. 100 mL/bottle. Store at room temperature.
SDP
Catalog No. NC1586996
Encompass
This item has a minimum order quantity of 96 per supplier requirements. MOLTOX manufactures a wide variety of buffers for your laboratory needs. All MOLTOX reagents are accompanied by a GLP-compliant Formulation and Quality Control statement.
Catalog No. NC1586996 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 26669
This item has a minimum order quantity of 96 per supplier requirements. MOLTOX manufactures a wide variety of buffers for your laboratory needs. All MOLTOX reagents are accompanied by a GLP-compliant Formulation and Quality Control statement.