Learn More
Teknova PPB Solution. 1000mL, Sterile.
SDP
Catalog No. 50842975
Encompass
30mM Tris-HCl pH 8.0, 1mM Ethylenediaminetetraacetic acid (EDTA) pH 8.0,20% Sucrose
Catalog No. 50-842-975 Supplier Teknova Supplier No. P2140
30mM Tris-HCl pH 8.0, 1mM Ethylenediaminetetraacetic acid (EDTA) pH 8.0,20% Sucrose