Learn More
Specifications
Specifications
| Cross Reactivity | Human |
| Description | PPBP (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human PPBP. |
| Detection Method | Colorimetric |
| Quantity | 1 Kit |
| Sample Type | Cell Lysates, Plasma, Serum |
| Sample Volume | 100 μL |
| Content And Storage | Store the kit at 4°C |
| For Use With (Application) | Quantification |
| Product Type | PPBP (Human) ELISA Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.