Learn More
Aviva Systems Biology Corp PPCS Antibody - C-terminal region (ARP63677_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63677_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPCS. It was validated on Western Blot. . Alternate names: FLJ11838, MGC117357, MGC138220, RP11-163G10.1ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-288-051