Learn More
Cayman Chemical Pipequaline, Purity: ≥98%, CAS: 77472-98-1, 10 mg
SDP
Catalog No. 502964412
Encompass_Preferred
A partial agonist of central benzodiazepine receptors (Ki = 78 nM in a radioligand binding assay); potentiates muscle relaxant and hypnotic effects of diazepam in mice; increases drinking in the Vogel punished drinking task indicating anxiolytic-like activity
Catalog No. 50-296-4412 Supplier Cayman Chemical Supplier No. 2388810 MG
A partial agonist of central benzodiazepine receptors (Ki = 78 nM in a radioligand binding assay); potentiates muscle relaxant and hypnotic effects of diazepam in mice; increases drinking in the Vogel punished drinking task indicating anxiolytic-like activity